2-(1-Benzyl-3-pyrrolidinyl)acetic acid
Supplier: Matrix Scientific
053976-500MG
053976-2.5G
101905-712EA
364.62
USD
101905-712
101905-714
2-(1-Benzyl-3-pyrrolidinyl)acetic acid
2-(1-Benzyl-3-pyrrolidinyl)acetic acid
Hazard: Irritant
Formula:
C₁₃H₁₇NO₂ |
MDL Number:
MFCD04116217 CAS Number: 666853-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...