2-(1-Isobutyl-3-pyrrolidinyl)acetic acid

Supplier: Matrix Scientific

053975-500MG 053975-2.5G
101905-708EA 364.62 USD
101905-708 101905-710
2-(1-Isobutyl-3-pyrrolidinyl)acetic acid
2-(1-Isobutyl-3-pyrrolidinyl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₉NO₂
MDL Number: MFCD13563020
CAS Number: 1220019-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR