2-(1-Isopropyl-3-pyrrolidinyl)acetic acid

Supplier: Matrix Scientific

053973-500MG 053973-2.5G
101905-700EA 461.54 USD
101905-700 101905-702
2-(1-Isopropyl-3-pyrrolidinyl)acetic acid
2-(1-Isopropyl-3-pyrrolidinyl)acetic acid

Hazard: Irritant

Formula: C₉H₁₇NO₂
MDL Number: MFCD13563018
CAS Number: 1220030-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR