2-(1-Methyl-3-pyrrolidinyl)acetic acid

Supplier: Matrix Scientific

053971-500MG 053971-2.5G
101905-692EA 592.31 USD
101905-692 101905-694
2-(1-Methyl-3-pyrrolidinyl)acetic acid
2-(1-Methyl-3-pyrrolidinyl)acetic acid

Hazard: Irritant

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD13563016
CAS Number: 102014-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR