Ethyl-2-(3-pyrrolidinylidene)acetate
Supplier: Matrix Scientific
053970-500MG
053970-2.5G
101905-688EA
364.62
USD
101905-688
101905-690
Ethyl-2-(3-pyrrolidinylidene)acetate
Ethyl-2-(3-pyrrolidinylidene)acetate
Hazard: Irritant
Formula:
C₈H₁₃NO₂ |
MDL Number:
MFCD28902405 CAS Number: 1220040-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...