2-(3-Pyrrolidinyl)acetic acid hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-Pyrrolidineacetic acid

053969-500MG 053969-2.5G
101905-684EA 364.62 USD
101905-684 101905-686
2-(3-Pyrrolidinyl)acetic acid hydrochloride
2-(3-Pyrrolidinyl)acetic acid hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
MDL Number: MFCD06795694
CAS Number: 89203-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR