N-Methyl-2-(1-methyl-3-piperidinyl)-1-ethanamine
Supplier: Matrix Scientific
053967-500MG
053967-2.5G
101905-676EA
166.15
USD
101905-676
101905-678
N-Methyl-2-(1-methyl-3-piperidinyl)-1-ethanamine
N-Methyl-2-(1-methyl-3-piperidinyl)-1-ethanamine
Hazard: Irritant
Formula:
C₉H₂₀N₂ |
MDL Number:
MFCD11847950 CAS Number: 807297-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...