2-(1-Methyl-3-piperidinyl)ethylamine

Supplier: Matrix Scientific

053966-500MG 053966-2.5G
101905-672EA 538.46 USD
101905-672 101905-674
2-(1-Methyl-3-piperidinyl)ethylamine
2-(1-Methyl-3-piperidinyl)ethylamine

Hazard: Irritant

Formula: C₈H₁₈N₂
MW: 142.25 g/mol
MDL Number: MFCD12024737
CAS Number: 53295-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR