2-(1-Methyl-3-piperidinyl)-1-ethanol

Supplier: Matrix Scientific

053965-500MG 053965-2.5G
101905-668EA 326.15 USD
101905-668 101905-670
2-(1-Methyl-3-piperidinyl)-1-ethanol
2-(1-Methyl-3-piperidinyl)-1-ethanol

Hazard: Irritant

Formula: C₈H₁₇NO
MW: 143.23 g/mol
Boiling Pt: 111…112 °C (5 Torr)
MDL Number: MFCD13183972
CAS Number: 101257-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR