2-(1-Benzyl-3-piperidinyl)acetic acid

Supplier: Matrix Scientific

053964-500MG 053964-2.5G
101905-664EA 669.23 USD
101905-664 101905-666
2-(1-Benzyl-3-piperidinyl)acetic acid
2-(1-Benzyl-3-piperidinyl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₂
MDL Number: MFCD11103713
CAS Number: 64995-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR