2-(1-Isobutyl-3-piperidinyl)acetic acid

Supplier: Matrix Scientific

053963-500MG 053963-2.5G
101905-660EA 364.62 USD
101905-660 101905-662
2-(1-Isobutyl-3-piperidinyl)acetic acid
2-(1-Isobutyl-3-piperidinyl)acetic acid

Hazard: Irritant

Formula: C₁₁H₂₁NO₂
MDL Number: MFCD13563013
CAS Number: 1220019-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR