2-(1-Isopropyl-3-piperidinyl)acetic acid
Supplier: Matrix Scientific
053962-500MG
053962-2.5G
101905-656EA
592.31
USD
101905-656
101905-658
2-(1-Isopropyl-3-piperidinyl)acetic acid
2-(1-Isopropyl-3-piperidinyl)acetic acid
Hazard: Irritant
Formula:
C₁₀H₁₉NO₂ |
MDL Number:
MFCD13563012 CAS Number: 1220030-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...