2-(1-Ethyl-3-piperidinyl)acetic acid

Supplier: Matrix Scientific

053961-500MG 053961-2.5G
101905-652EA 364.62 USD
101905-652 101905-654
2-(1-Ethyl-3-piperidinyl)acetic acid
2-(1-Ethyl-3-piperidinyl)acetic acid

Hazard: Irritant

Formula: C₉H₁₇NO₂
MDL Number: MFCD13563011
CAS Number: 1220038-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR