Ethyl-2-(3-piperidinyl)acetate

Supplier: Matrix Scientific

Synonyms: Ethyl (piperidin-3-yl)acetate

053959-500MG 053959-2.5G
101905-644EA 364.62 USD
101905-644 101905-646
Ethyl-2-(3-piperidinyl)acetate
Ethyl-2-(3-piperidinyl)acetate

Hazard: Irritant

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
MDL Number: MFCD03412125
CAS Number: 64995-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR