Ethyl-2-(3-piperidinylidene)acetate

Supplier: Matrix Scientific

053958-500MG 053958-2.5G
101905-640EA 364.62 USD
101905-640 101905-642
Ethyl-2-(3-piperidinylidene)acetate
Ethyl-2-(3-piperidinylidene)acetate

Hazard: Irritant

Formula: C₉H₁₅NO₂
MDL Number: MFCD13563010
CAS Number: 957752-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR