4-(4-Methylbenzyl)-1,4-diazepan-5-one

Supplier: Matrix Scientific

053956-500MG 053956-2.5G
101905-632EA 364.62 USD
101905-632 101905-634
4-(4-Methylbenzyl)-1,4-diazepan-5-one
4-(4-Methylbenzyl)-1,4-diazepan-5-one

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MW: 218.3 g/mol
MDL Number: MFCD13563008
CAS Number: 1220038-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR