4-(2-Methylbenzyl)-1,4-diazepan-5-one

Supplier: Matrix Scientific

053954-500MG 053954-2.5G
101905-624EA 364.62 USD
101905-624 101905-626
4-(2-Methylbenzyl)-1,4-diazepan-5-one
4-(2-Methylbenzyl)-1,4-diazepan-5-one

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MDL Number: MFCD13563006
CAS Number: 1220036-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR