4-(2-Methoxybenzyl)-1,4-diazepan-5-one

Supplier: Matrix Scientific

053952-500MG 053952-2.5G
101905-616EA 149.23 USD
101905-616 101905-618
4-(2-Methoxybenzyl)-1,4-diazepan-5-one
4-(2-Methoxybenzyl)-1,4-diazepan-5-one

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD13563004
CAS Number: 1220030-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR