4-(3-Methoxybenzyl)-1,4-diazepan-5-one
Supplier: Matrix Scientific
053950-500MG
053950-2.5G
101905-608EA
461.54
USD
101905-608
101905-610
4-(3-Methoxybenzyl)-1,4-diazepan-5-one
4-(3-Methoxybenzyl)-1,4-diazepan-5-one
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₂ MW: 234.3 g/mol |
MDL Number:
MFCD13563002 CAS Number: 1220036-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...