4-(4-Fluorobenzyl)-1,4-diazepan-5-one

Supplier: Matrix Scientific

053948-500MG 053948-2.5G
101905-600EA 364.62 USD
101905-600 101905-602
4-(4-Fluorobenzyl)-1,4-diazepan-5-one
4-(4-Fluorobenzyl)-1,4-diazepan-5-one

Hazard: Irritant/Stored under Argon

Formula: C₁₂H₁₅FN₂O
MW: 222.26 g/mol
MDL Number: MFCD13563001
CAS Number: 1220035-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR