4-(Cyclopropylmethyl)-1,4-diazepan-5-one hydrochloride

Supplier: Matrix Scientific

053945-500MG 053945-2.5G
101905-588EA 461.54 USD
101905-588 101905-590
4-(Cyclopropylmethyl)-1,4-diazepan-5-one hydrochloride
4-(Cyclopropylmethyl)-1,4-diazepan-5-one hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O
MDL Number: MFCD22421954
CAS Number: 1401425-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR