4-Benzyl-1,4-diazepan-5-one hydrochloride

Supplier: Matrix Scientific

053941-500MG 053941-2.5G
101905-572EA 538.46 USD
101905-572 101905-574
4-Benzyl-1,4-diazepan-5-one hydrochloride
4-Benzyl-1,4-diazepan-5-one hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MW: 240.74 g/mol
MDL Number: MFCD18459278
CAS Number: 1211513-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR