4-Propyl-1,4-diazepan-5-one hydrochloride

Supplier: Matrix Scientific

053940-500MG 053940-2.5G
101905-568EA 461.54 USD
101905-568 101905-570
4-Propyl-1,4-diazepan-5-one hydrochloride
4-Propyl-1,4-diazepan-5-one hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD16628200
CAS Number: 1241675-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR