4-Methyl-1,4-diazepan-5-one hydrochloride

Supplier: Matrix Scientific

053938-500MG 053938-2.5G
101905-560EA 384.62 USD
101905-560 101905-562
4-Methyl-1,4-diazepan-5-one hydrochloride
4-Methyl-1,4-diazepan-5-one hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD16872041
CAS Number: 329794-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR