4-Amino-1-(2-ethoxyethyl)-2-pyrrolidinone

Supplier: Matrix Scientific

053937-500MG 053937-2.5G
101905-556EA 167.69 USD
101905-556 101905-558
4-Amino-1-(2-ethoxyethyl)-2-pyrrolidinone
4-Amino-1-(2-ethoxyethyl)-2-pyrrolidinone

Hazard: Irritant

Formula: C₈H₁₆N₂O₂
MDL Number: MFCD12080226
CAS Number: 1155146-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR