4-Amino-1-propyl-2-pyrrolidinone
Supplier: Matrix Scientific
053935-500MG
053935-2.5G
101905-548EA
167.69
USD
101905-548
101905-550
4-Amino-1-propyl-2-pyrrolidinone
4-Amino-1-propyl-2-pyrrolidinone
Hazard: Irritant
Formula:
C₇H₁₄N₂O |
MDL Number:
MFCD09055321 CAS Number: 1083245-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...