2-(2-Thienyl)azetidine

Supplier: Matrix Scientific

053931-500MG 053931-2.5G
101905-536EA 364.62 USD
101905-536 101905-538
2-(2-Thienyl)azetidine
2-(2-Thienyl)azetidine

Hazard: Irritant

Formula: C₇H₉NS
MW: 139.22 g/mol
MDL Number: MFCD06217577
CAS Number: 777886-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR