1-(3-Aminopropyl)-3-azetidinol

Supplier: Matrix Scientific

053929-500MG 053929-2.5G
101905-528EA 384.62 USD
101905-528 101905-530
1-(3-Aminopropyl)-3-azetidinol
1-(3-Aminopropyl)-3-azetidinol

Hazard: Irritant

Formula: C₆H₁₄N₂O
MDL Number: MFCD09745963
CAS Number: 1219979-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR