(3-Amino-1-azetidinyl)(phenyl)methanone

Supplier: Matrix Scientific

053926-500MG 053926-2.5G
101905-516EA 364.62 USD
101905-516 101905-518
(3-Amino-1-azetidinyl)(phenyl)methanone
(3-Amino-1-azetidinyl)(phenyl)methanone

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD07786941
CAS Number: 887588-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR