N-(3-Azetidinyl)-3-methoxybenzamide hydrochloride

Supplier: Matrix Scientific

053925-500MG 053925-2.5G
101905-512EA 364.62 USD
101905-512 101905-514
N-(3-Azetidinyl)-3-methoxybenzamide hydrochloride
N-(3-Azetidinyl)-3-methoxybenzamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₂
MDL Number: MFCD22421953
CAS Number: 1401425-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR