N-(3-Azetidinyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

053924-500MG 053924-2.5G
101905-508EA 364.62 USD
101905-508 101905-510
N-(3-Azetidinyl)cyclobutanecarboxamide
N-(3-Azetidinyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₈H₁₄N₂O
MDL Number: MFCD13562988
CAS Number: 1220031-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR