N-(3-Azetidinyl)cyclopentanecarboxamide

Supplier: Matrix Scientific

053923-500MG 053923-2.5G
101905-504EA 364.62 USD
101905-504 101905-506
N-(3-Azetidinyl)cyclopentanecarboxamide
N-(3-Azetidinyl)cyclopentanecarboxamide

Hazard: Irritant

Formula: C₉H₁₆N₂O
MDL Number: MFCD13562987
CAS Number: 1220027-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR