1-(3-Azetidinyl)piperidine

Supplier: Matrix Scientific

053920-500MG 053920-2.5G
101905-492EA 423.08 USD
101905-492 101905-494
1-(3-Azetidinyl)piperidine
1-(3-Azetidinyl)piperidine

Hazard: Irritant

Formula: C₈H₁₆N₂
MDL Number: MFCD09264396
CAS Number: 138022-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR