1-(3-Azetidinyl)pyrrolidine dihydrochloride

Supplier: Matrix Scientific

053919-500MG 053919-2.5G
101905-488EA 384.62 USD
101905-488 101905-490
1-(3-Azetidinyl)pyrrolidine dihydrochloride
1-(3-Azetidinyl)pyrrolidine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₆Cl₂N₂
MW: 199.12 g/mol
MDL Number: MFCD11617821
CAS Number: 149105-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR