N,N-Diethyl-3-azetidinamine dihydrochloride

Supplier: Matrix Scientific

053918-500MG 053918-2.5G
101905-484EA 538.46 USD
101905-484 101905-486
N,N-Diethyl-3-azetidinamine dihydrochloride
N,N-Diethyl-3-azetidinamine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₈Cl₂N₂
MW: 201.14 g/mol
MDL Number: MFCD18381080
CAS Number: 55438-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR