1-(3-Methoxybenzyl)-4-oxo-2-azetidine carboxylicacid

Supplier: Matrix Scientific

053916-500MG 053916-2.5G
101905-476EA 364.62 USD
101905-476 101905-478
1-(3-Methoxybenzyl)-4-oxo-2-azetidine carboxylicacid
1-(3-Methoxybenzyl)-4-oxo-2-azetidine carboxylicacid

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
MDL Number: MFCD13562985
CAS Number: 1236267-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR