1-Benzyl-4-oxo-2-azetidinecarboxylic acid

Supplier: Matrix Scientific

053915-500MG 053915-2.5G
101905-472EA 364.62 USD
101905-472 101905-474
1-Benzyl-4-oxo-2-azetidinecarboxylic acid
1-Benzyl-4-oxo-2-azetidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD13562984
CAS Number: 1236267-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR