4-Oxo-1-propyl-2-azetidinecarboxylic acid

Supplier: Matrix Scientific

053914-500MG 053914-2.5G
101905-468EA 364.62 USD
101905-468 101905-470
4-Oxo-1-propyl-2-azetidinecarboxylic acid
4-Oxo-1-propyl-2-azetidinecarboxylic acid

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD13562983
CAS Number: 1246172-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR