2-(4-Methyl-3-oxo-1-propyl-2-piperazinyl)acetic acid

Supplier: Matrix Scientific

053898-500MG 053898-2.5G
101905-404EA 364.62 USD
101905-404 101905-406
2-(4-Methyl-3-oxo-1-propyl-2-piperazinyl)acetic acid
2-(4-Methyl-3-oxo-1-propyl-2-piperazinyl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₈N₂O₃
MDL Number: MFCD06248291
CAS Number: 1236257-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR