2-(1-Benzyl-4-methyl-3-oxo-2-piperazinyl)acetic acid

Supplier: Matrix Scientific

053897-500MG 053897-2.5G
101905-400EA 364.62 USD
101905-400 101905-402
2-(1-Benzyl-4-methyl-3-oxo-2-piperazinyl)acetic acid
2-(1-Benzyl-4-methyl-3-oxo-2-piperazinyl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃
MDL Number: MFCD06245861
CAS Number: 1236267-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR