2-(1-Ethyl-4-methyl-3-oxo-2-piperazinyl)acetic acid

Supplier: Matrix Scientific

053895-500MG 053895-2.5G
101905-392EA 364.62 USD
101905-392 101905-394
2-(1-Ethyl-4-methyl-3-oxo-2-piperazinyl)acetic acid
2-(1-Ethyl-4-methyl-3-oxo-2-piperazinyl)acetic acid

Hazard: Irritant

Formula: C₉H₁₆N₂O₃
MDL Number: MFCD13562968
CAS Number: 1236261-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR