Methyl-2-(4-benzyl-3-oxo-2-piperazinyl)acetate

Supplier: Matrix Scientific

053893-500MG 053893-2.5G
101905-384EA 461.54 USD
101905-384 101905-386
Methyl-2-(4-benzyl-3-oxo-2-piperazinyl)acetate
Methyl-2-(4-benzyl-3-oxo-2-piperazinyl)acetate

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃
MDL Number: MFCD08689435
CAS Number: 534603-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR