Methyl-2-(4-ethyl-3-oxo-2-piperazinyl)acetate

Supplier: Matrix Scientific

053892-500MG 053892-2.5G
101905-380EA 364.62 USD
101905-380 101905-382
Methyl-2-(4-ethyl-3-oxo-2-piperazinyl)acetate
Methyl-2-(4-ethyl-3-oxo-2-piperazinyl)acetate

Hazard: Irritant

Formula: C₉H₁₆N₂O₃
MDL Number: MFCD13562966
CAS Number: 534603-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR