(4-Methyl-3-pyridinyl)methanamine

Supplier: Matrix Scientific

Synonyms: (4-Methylpyridin-3-yl)methanamine

053890-500MG 053890-2.5G
101905-372EA 364.62 USD
101905-372 101905-374
(4-Methyl-3-pyridinyl)methanamine
(4-Methyl-3-pyridinyl)methanamine

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD06213859
CAS Number: 1443-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR