(6-Methylpyridin-2-yl)methanamine

Supplier: Matrix Scientific

053889-500MG 053889-2.5G
101905-368EA 95.38 USD
101905-368 101905-370
(6-Methylpyridin-2-yl)methanamine
(6-Methylpyridin-2-yl)methanamine

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.1700 g/mol
MDL Number: MFCD05864530
CAS Number: 6627-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR