N-[(3-Methyl-2-pyridinyl)methyl]-1-ethanamine
Supplier: Matrix Scientific
053887-500MG
053887-2.5G
101905-360EA
364.62
USD
101905-360
101905-362
N-[(3-Methyl-2-pyridinyl)methyl]-1-ethanamine
N-[(3-Methyl-2-pyridinyl)methyl]-1-ethanamine
Hazard: Irritant
Formula:
C₉H₁₄N₂ MW: 150.23 g/mol |
MDL Number:
MFCD13562964 CAS Number: 952194-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...