(3-Methyl-2-pyridinyl)methanamine

Supplier: Matrix Scientific

Synonyms: 2-(Aminomethyl)-3-methylpyridine

053885-500MG 053885-2.5G
101905-352EA 100 USD
101905-352 101905-354
(3-Methyl-2-pyridinyl)methanamine
(3-Methyl-2-pyridinyl)methanamine

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD04116209
CAS Number: 153936-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR