2-(4-Pyridinyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol
Supplier: Matrix Scientific
053883-500MG
053883-2.5G
101905-344EA
364.62
USD
101905-344
101905-346
2-(4-Pyridinyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol
2-(4-Pyridinyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol
Hazard: Irritant
Formula:
C₁₁H₁₀N₄O |
MDL Number:
MFCD13562962 CAS Number: 1220031-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...