2-(2-Pyridinyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol
Supplier: Matrix Scientific
053882-500MG
053882-2.5G
101905-340EA
364.62
USD
101905-340
101905-342
2-(2-Pyridinyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol
2-(2-Pyridinyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol
Hazard: Irritant
Formula:
C₁₁H₁₀N₄O |
MDL Number:
MFCD13562961 CAS Number: 1220031-07-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...