1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride

Supplier: Matrix Scientific

Synonyms: 1,4,5,6-Tetrahydro-pyrrolo[3,4-c]pyrazole,hydrochloride

053874-500MG 053874-2.5G
101905-308EA 206.15 USD
101905-308 101905-310
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride

Hazard: Irritant

Formula: C₅H₉Cl₂N₃
MDL Number: MFCD08457551
CAS Number: 157327-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR